Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Fas-Associated protein with Death Domain
- Antigen Symbol:FADD
- Clonality:Monoclonal
- Clone:J1D2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:8772
- Antigen Synonyms:FAS-associating death domain-containing protein|MORT1Fas-associating protein with death domain|Growth-inhibiting gene 3 protein|mediator of receptor-induced toxicity|Fas (TNFRSF6)-associated via death domain|protein FADD|MGC8528|GIG3|FAS-associated death domain protein|Mediator of receptor induced toxicity
- Storage Buffer:PBS (pH 7.4) and 10% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:1.0 mg/ml
- Immunogen:Recombinant human FADD (1-208aa) purified from E. coli
- Purification:Protein G purified
- Cat. No.:102156-068
- Supplier no.:NBP1-04288
Specifications
About this item
The FADD Antibody (J1D2) from Novus Biologicals is a mouse monoclonal antibody to FADD. This antibody reacts with human. The FADD Antibody (J1D2) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:50
Type: Primary
Antigen: FADD
Clonality: Monoclonal
Clone: J1D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human