Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Estrogen Receptor 1
- Antigen Symbol:ER alpha/NR3A1
- Clonality:Monoclonal
- Clone:C-542
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Hamster,Human,Rat,Mouse
- Western Blot:Yes
- Size:0.05 mg
- Gene ID:2099
- Antigen Synonyms:Estradiol receptor|ER|estrogen receptor alpha|Era|NR3A1DKFZp686N23123|ER alpha|estrogen receptor alpha delta 4*5,6,7*/654 isoform|ESRESRA|estrogen receptor|ER-alpha|estrogen receptor 1|Nuclear receptor subfamily 3 group A member 1|estrogen receptor alpha delta 3*4,56,7*/819-2 isoform|estrogen receptor alpha delta 4 +49 isoform
- Storage Buffer:PBS and 50% Glycerol
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:Synthetic peptide corresponding to the sequence near the C-terminus of human Estrogen Receptor alpha.
- Purification:Protein G purified
- Cat. No.:102158-672
- Supplier no.:NBP1-19318
Specifications
About this item
The ER alpha / NR3A1 Antibody (C-542) from Novus Biologicals is a mouse monoclonal antibody to ER alpha / NR3A1. This antibody reacts with human, mouse, rat, hamster. The ER alpha / NR3A1 Antibody (C-542) has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: ER alpha/NR3A1
Clonality: Monoclonal
Clone: C-542
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Hamster