Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:D-Dimer
- Antigen Symbol:D-Dimer
- Clonality:Monoclonal
- Clone:DD41
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Western Blot:Yes
- Size:0.2 mg
- Antigen Synonyms:D Dimer
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Store at 4C. Do not freeze.
- Immunogen:Derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with homogenized fibrin clot, D-dimer or high molecular weight fibrin degradation products.
- Purification:Protein A purified
- Cat. No.:102115-856
- Supplier no.:NB100-73038
Specifications
About this item
The D-Dimer Antibody (DD41) from Novus Biologicals is a mouse monoclonal antibody to D-Dimer. This antibody reacts with human. The D-Dimer Antibody (DD41) has been validated for the following applications: Western Blot, Sandwich ELISA.
Type: Primary
Antigen: D-Dimer
Clonality: Monoclonal
Clone: DD41
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human