Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Symbol:CNTFR
- Clonality:Monoclonal
- Clone:MM0190-10L29
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Form:Lyophilized
- Gene ID:1271
- Antigen Synonyms:CNTF receptor subunit alpha|CNTFR alpha|ciliary neurotrophic factor receptor alpha|CNTFR-alpha|MGC1774|ciliary neurotrophic factor receptor|ciliary neurotrophic factor receptor subunit alpha
- Storage Buffer:A 0.2 um filtered solution in PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:Human CNTFR
- Purification:Protein G purified
- Cat. No.:102251-128
- Supplier no.:NBP2-12196
Specifications
About this item
The CNTFR Antibody (MM0190-10L29) from Novus Biologicals is a mouse monoclonal antibody to CNTFR. This antibody reacts with human. The CNTFR Antibody (MM0190-10L29) has been validated for the following applications: Western Blot, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: CNTFR
Clonality: Monoclonal
Clone: MM0190-10L29
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human