About this item
The c-Myc Antibody (9E10) from Novus Biologicals is a mouse monoclonal antibody to c-Myc. This antibody reacts with human, mouse, drosophila. The c-Myc Antibody (9E10) has been validated for the following applications: Western Blot, Simple Western, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Type: Primary
Antigen: c-Myc
Clonality: Monoclonal
Clone: 9E10
Conjugation: Unconjugated
Epitope: Leucine zipper sequence (AEEQKLISEEDLL)
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Drosophila
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Catalog No:
- 102133-696
- 102133-708
- Antigen Symbol:
- c-Myc
- c-Myc
- Antigen Synonyms:
- v-myc avian myelocytomatosis viral oncogene homolog|Class E basic helix-loop-helix protein 39|v-myc myelocytomatosis viral oncogene homolog (avian)|avian myelocytomatosis viral oncogene homolog|9E10 anti-Myc|9E10 Myc|myc proto-oncogene protein|Proto-oncogene c-Myc|BHLHE39|myc-related translation/localization regulatory factor|MYC|c-Myc|Transcription factor p64|bHLHe39MRTL|MYCC
- v-myc avian myelocytomatosis viral oncogene homolog|Class E basic helix-loop-helix protein 39|v-myc myelocytomatosis viral oncogene homolog (avian)|avian myelocytomatosis viral oncogene homolog|9E10 anti-Myc|9E10 Myc|myc proto-oncogene protein|Proto-oncogene c-Myc|BHLHE39|myc-related translation/localization regulatory factor|MYC|c-Myc|Transcription factor p64|bHLHe39MRTL|MYCC
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 9E10
- 9E10
- Reactivity:
- Drosophila, Human, Mouse
- Drosophila, Human, Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- 4609
- 4609
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Epitope:
- Leucine zipper sequence (AEEQKLISEEDLL)
- Leucine zipper sequence (AEEQKLISEEDLL)
- Immunogen:
- A synthetic peptide corresponding to amino acids 408-439 (AEEQKLISEEDLLRKRREQLKHKLEQLRNSCA) of human c-Myc. [UniProt# P01106]
- A synthetic peptide corresponding to amino acids 408-439 (AEEQKLISEEDLLRKRREQLKHKLEQLRNSCA) of human c-Myc. [UniProt# P01106]
- Purification:
- Protein G purified
- Protein G purified
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Storage Buffer:
- Tris-Glycine and 0.15M NaCl
- Tris-Glycine and 0.15M NaCl
- Storage Temperature:
- Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.