To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:CD8 alpha
- Clonality:Monoclonal
- Clone:4B11
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b
- Reactivity:Human
- Size:100 μl
- Gene ID:925
- Antigen Synonyms:T8 T-cell antigen|p32|OKT8 T-cell antigen|T-cell antigen Leu2|RPA-T8|RPA-T8 Flow|alpha polypeptide (p32)|T-lymphocyte differentiation antigen T8/Leu-2|RPA-T8 antibody flow|Leu2 T-lymphocyte antigen|T-cell surface glycoprotein CD8 alpha chain|RPA-T8 Clone|T cell co-receptor|CD8 antigen|Leu2|MAL|CD8|RPA-T8 CD8|CD8a antigen|CD8a molecule
- Storage Buffer:No buffer
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:Synthetic peptide derived from the carboxy terminal region of the humanCD8 alpha chain coupled to a N-terminal cysteine, with the sequence C-KSDGKPSLSARYV. The peptide was coupled to bovine serum albuman and keyhole limpet hemocyanin.
- Purification:Tissue culture supernatant
- Cat. No.:102114-970
- Supplier no.:NB100-65729
Specifications
About this item
The CD8 alpha Antibody (4B11) from Novus Biologicals is a mouse monoclonal antibody to CD8 alpha. This antibody reacts with human. The CD8 alpha Antibody (4B11) has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:20 - 1:40
Type: Primary
Antigen: CD8 alpha
Clonality: Monoclonal
Clone: 4B11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human