Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:CD4 molecule
- Antigen Symbol:CD4
- Clonality:Monoclonal
- Clone:CT-4
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Turkey,Birds,Chicken
- Size:0.25 mg
- Gene ID:920
- Antigen Synonyms:CD4 antigen (p55)|CD4 antigen|CD4 receptor|T-cell surface antigen T4/Leu-3|CD4mut|T-cell surface glycoprotein CD4|CD4 molecule
- Storage Buffer:PBS
- Storage Temperature:Store at 4C. Do not freeze.
- Immunogen:Chicken thymocytes and Ig-negative blood leukocytes
- Purification:Protein A or G purified
- Cat. No.:102161-850
- Supplier no.:NBP1-28267
The CD4 Antibody (CT-4) [FITC] from Novus Biologicals is a mouse monoclonal antibody to CD4. This antibody reacts with avian, chicken, turkey. The CD4 Antibody (CT-4) [FITC] has been validated for the following applications: Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: CT-4
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Avian, Chicken, Turkey