Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Butyrophilin
- Antigen Symbol:CD277/BTN3A1
- Clonality:Monoclonal
- Clone:BT3.1
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Size:100 μg
- Gene ID:11119
- Storage Buffer:PBS, pH 7.2
- Storage Temperature:Store at 4C. Do not freeze.
- Concentration:0.5 mg/ml
- Immunogen:The immunogen for this antibody is CD277 (B7 family, Butyrophilin).
- Purification:Protein A or G purified
- Cat. No.:102227-006
- Supplier no.:NBP2-00227
Specifications
About this item
The CD277 / BTN3A1 Antibody (BT3.1) from Novus Biologicals is a mouse monoclonal antibody to CD277 / BTN3A1. This antibody reacts with human. The CD277 / BTN3A1 Antibody (BT3.1) has been validated for the following applications: Flow Cytometry, Immunoprecipitation.
Type: Primary
Antigen: CD277/BTN3A1
Clonality: Monoclonal
Clone: BT3.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human