Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:Basigin
- Antigen Symbol:EMMPRIN/CD147
- Clonality:Monoclonal
- Clone:MEM-M6/1
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Horse,Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:682
- Antigen Synonyms:CD147|EMMPRINTCSF|Tumor cell-derived collagenase stimulatory factor|basigin (Ok blood group)|basigin|Leukocyte activation antigen M6|Extracellular matrix metalloproteinase inducer|Collagenase stimulatory factor|5F7|OK blood group antigen|OK|CD147 antigen|M6
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Store at 4C. Do not freeze.
- Concentration:1.0 mg/ml
- Immunogen:Protein A-CR purified soluble recombinant form of CD147, CD147Rg, which consists of the cDNA coding for the hinge region, C2-and C3 domain of human IgG1, CD147Rg is secreted by transfectants as a dimer (NM_001728.2).
- Purification:Protein A purified
- Cat. No.:102132-446
- Supplier no.:NB500-430
The EMMPRIN / CD147 Antibody (MEM-M6 / 1) from Novus Biologicals is a mouse monoclonal antibody to EMMPRIN / CD147. This antibody reacts with human, equine. The EMMPRIN / CD147 Antibody (MEM-M6 / 1) has been validated for the following applications: Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, CyTOF-ready.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:200
Type: Primary
Antigen: EMMPRIN/CD147
Clonality: Monoclonal
Clone: MEM-M6/1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Equine