Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Integrin alpha L
- Antigen Symbol:ITGAL
- Clonality:Monoclonal
- Clone:MEM-83
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Size:100 μg
- Gene ID:3683
- Antigen Synonyms:LFA-1 alpha|LFA-1|alpha L (antigen CD11A (p180)|alphapolypeptide|CD11a antigen|alpha polypeptide)|lymphocyte function-associated antigen1|Leukocyte adhesion glycoprotein LFA-1 alpha chain|LFA1A|Leukocyte function-associated molecule 1 alpha chain|CD11Aantigen CD11A (p180)|integrin gene promoter|CD11a|CD11 antigen-like family member A|integrin alpha-L|LFA-1A|lymphocyte function-associated antigen 1|integrin
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Store at 4C. Do not freeze.
- Concentration:1.0 mg/ml
- Immunogen:Human peripheral blood lymphocytes
- Purification:Protein A purified
- Cat. No.:102132-216
- Supplier no.:NB500-309
Specifications
About this item
Mouse Monoclonal CD11a Antibody (MEM-83). Tested Applications: Flow Cytometry, Functional, Immunoprecipitation. Tested Reactivity: Human.
Type: Primary
Antigen: CD11a
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human