Order Entry
United States
ContactUsLinkComponent
Anti-CD1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 76-7-4]
Anti-CD1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 76-7-4]
Catalog # 102161-766
Supplier:  Novus Biologicals
Anti-CD1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 76-7-4]
Catalog # 102161-766
Supplier:  Novus Biologicals
Supplier Number:  NBP1-28222
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Symbol:
    CD1
  • Clonality:
    Monoclonal
  • Clone:
    76-7-4
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Pig
  • Size:
    0.25 mg
  • Gene ID:
    909
  • Antigen Synonyms:
    FCB6|epidermal dendritic cell marker CD1a|cortical thymocyte antigen CD1A|T-cell surface antigen T6/Leu-6|cluster of differentiation 1 A|T6|CD1|R4|CD1a molecule|a polypeptide|T-cell surface glycoprotein CD1a|CD1a antigen|differentiation antigen CD1-alpha-3|CD1A antigen|hTa1 thymocyte antigen|HTA1
  • Storage Buffer:
    PBS
  • Storage Temperature:
    Store at 4C. Do not freeze.
  • Immunogen:
    Fresh dd miniature swine thymocytes
  • Purification:
    Protein A or G purified
  • Cat. No.:
    102161-766
  • Supplier no.:
    NBP1-28222

Specifications

About this item

The CD1 Antibody (76-7-4) [FITC] from Novus Biologicals is a mouse monoclonal antibody to CD1. This antibody reacts with porcine. The CD1 Antibody (76-7-4) [FITC] has been validated for the following applications: Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen, Immunocytochemistry.

Type: Primary
Antigen: CD1
Clonality: Monoclonal
Clone: 76-7-4
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Porcine