Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:Complement Component C1qA
- Antigen Symbol:C1qA
- Clonality:Monoclonal
- Clone:JL-1
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2b
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:0.05 mg
- Gene ID:712
- Antigen Synonyms:A chain|q subcomponent|C1QA complement component 1
- Storage Buffer:0.2 um filtered solution in PBS and 0.1% BSA
- Storage Temperature:Store at 4C. Do not freeze.
- Concentration:0.1 mg/ml
- Immunogen:Purified mouse C1QA
- Purification:Protein A or G purified
- Cat. No.:102181-066
- Supplier no.:NBP1-51139
The Complement Component C1qA Antibody (JL-1) from Novus Biologicals is a mouse monoclonal antibody to Complement Component C1qA. This antibody reacts with human, rat. The Complement Component C1qA Antibody (JL-1) has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Frozen, Functional, Immunoassay.
Type: Primary
Antigen: C1qA
Clonality: Monoclonal
Clone: JL-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Rat