Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:beta-Actin
- Antigen Symbol:ACTB
- Clonality:Monoclonal
- Clone:8H10D10
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2b
- Reactivity:Primates,Chinese hamster,Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μl
- Gene ID:60
- Antigen Synonyms:PS1TP5-binding protein 1|cytoplasmic 1|PS1TP5BP1|ACTB|Beta-actin|beta|BRWS1|beta cytoskeletal actin|actin|Beta Actin
- Storage Buffer:PBS
- Storage Temperature:Store at 4 °C short term. Aliquot and store at −20 °C long term. Avoid freeze-thaw cycles.
- Concentration:1.0 mg/ml
- Immunogen:Synthetic peptide corresponding to amino-terminal residues of human Beta Actin, conjugated to KLH. [UniProt# P60709]
- Purification:Ammonium sulfate precipitation
- Cat. No.:102177-200
- Supplier no.:NBP1-47423
Specifications
About this item
The beta-Actin Antibody (8H10D10) from Novus Biologicals is a mouse monoclonal antibody to beta-Actin. This antibody reacts with human, mouse, rat, chinese hamster, primate. The beta-Actin Antibody (8H10D10) has been validated for the following applications: Western Blot, Simple Western, Flow Cytometry, ELISA, Immunocytochemistry / Immunofluorescence.
Simple Western
Type: Primary
Antigen: beta-Actin
Clonality: Monoclonal
Clone: 8H10D10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat, Chinese Hamster, Primate