About this item
The Apolipoprotein A5 Antibody (1G5G9) from Novus Biologicals is a mouse monoclonal antibody to Apolipoprotein A5. This antibody reacts with human. The Apolipoprotein A5 Antibody (1G5G9) has been validated for the following applications: Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, ICC / IF (Negative), CyTOF-ready.
Type: Primary
Antigen: APOAV
Clonality: Monoclonal
Clone: 1G5G9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Catalog No:
- 102122-636
- 102122-624
- Antigen Symbol:
- APOAV
- APOAV
- Antigen Name:
- Apolipoprotein A5
- Apolipoprotein A5
- Antigen Synonyms:
- apo-AV|MGC126836|FLJ97995|Apolipoprotein A5|Regeneration-associated protein 3|APOAV|APOA-V|apolipoprotein A-V|RAP3MGC126838
- apo-AV|MGC126836|FLJ97995|Apolipoprotein A5|Regeneration-associated protein 3|APOAV|APOA-V|apolipoprotein A-V|RAP3MGC126838
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 1G5G9
- 1G5G9
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 116519
- 116519
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- Ni-NTA purified full-length recombinant human Apolipoprotein A5 expressed in E. Coli strain BL21 (DE3). [UniProt# Q6Q788]
- Ni-NTA purified full-length recombinant human Apolipoprotein A5 expressed in E. Coli strain BL21 (DE3). [UniProt# Q6Q788]
- Purification:
- Protein G purified
- Protein G purified
- Concentration:
- 1.9 mg/ml
- 1.9 mg/ml
- Storage Buffer:
- PBS (pH 7.4)
- PBS (pH 7.4)
- Storage Temperature:
- Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.