Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:1-Acylglycerol-3-Phosphate O-Acyltransferase 5
- Antigen Symbol:AGPAT5
- Clonality:Monoclonal
- Clone:2C4
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:55326
- Antigen Synonyms:epsilon)|1AGPAT5,1-AGP acyltransferase 5|LPAATE|1-AGPAT 5,1-acylglycerol-3-phosphate O-acyltransferase 5|LPAAT-epsilon|EC 2.3.1.51|1-acyl-sn-glycerol-3-phosphate acyltransferase epsilon|Lysophosphatidic acid acyltransferase epsilon|FLJ11210|1-acylglycerol-3-phosphate O-acyltransferase 5 (lysophosphatidic acidacyltransferase|LPAAT-e
- Storage Buffer:PBS (pH 7.3), 1.0% BSA and 50% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:Full length human recombinant protein of human AGPAT5(NP_060831) producedin HEK293T cell.
- Purification:Immunogen affinity purified
- Cat. No.:102230-064
- Supplier no.:NBP2-01664
Specifications
About this item
The AGPAT5 Antibody (2C4) from Novus Biologicals is a mouse monoclonal antibody to AGPAT5. This antibody reacts with human. The AGPAT5 Antibody (2C4) has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: AGPAT5
Clonality: Monoclonal
Clone: 2C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human