Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:ADAM Metallopeptidase Domain 10
- Antigen Symbol:ADAM10
- Clonality:Monoclonal
- Clone:MM0077-6D31
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Form:Lyophilized
- Gene ID:102
- Antigen Synonyms:CDw156|ADAM metallopeptidase domain 10|CD156c|HsT18717|Mammalian disintegrin-metalloprotease|a disintegrin and metalloproteinase domain 10|a disintegrin and metalloprotease domain 10|Kuzbanian protein homolog|kuz|EC 3.4.24|CD156c antigen|ADAM 10|disintegrin and metalloproteinase domain-containing protein 10|AD10|MADMEC 3.4.24.81
- Storage Buffer:A 0.2 um filtered solution in PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:Human ADAM-10 (extracellular domain) recombinant protein.
- Purification:Protein G purified
- Cat. No.:102250-764
- Supplier no.:NBP2-12014
The ADAM10 Antibody (MM0077-6D31) from Novus Biologicals is a mouse monoclonal antibody to ADAM10. This antibody reacts with human. The ADAM10 Antibody (MM0077-6D31) has been validated for the following applications: Western Blot, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: ADAM10
Clonality: Monoclonal
Clone: MM0077-6D31
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human