About this item
The ABCG8 Antibody (1B10A5) from Novus Biologicals is a mouse monoclonal antibody to ABCG8. This antibody reacts with human, mouse. The ABCG8 Antibody (1B10A5) has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:100
Type: Primary
Antigen: ABCG8
Clonality: Monoclonal
Clone: 1B10A5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 102201-420
- 102201-418
- Antigen Symbol:
- ABCG8
- ABCG8
- Antigen Name:
- ATP Binding Cassette, sub-family G, member 8
- ATP Binding Cassette, sub-family G, member 8
- Antigen Synonyms:
- member 8|STSL|member 8 (sterolin 2)|sub-family G (WHITE)|ATP-binding cassette sub-family G member 8|subfamily G|sterolin 2|GBD4ATP-binding cassette|MGC142217|ATP-binding cassette|sterolin-2
- member 8|STSL|member 8 (sterolin 2)|sub-family G (WHITE)|ATP-binding cassette sub-family G member 8|subfamily G|sterolin 2|GBD4ATP-binding cassette|MGC142217|ATP-binding cassette|sterolin-2
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 1B10A5
- 1B10A5
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- 64241
- 64241
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Full length mouse ABCG8 [Swiss-Prot# Q9DBM0]
- Full length mouse ABCG8 [Swiss-Prot# Q9DBM0]
- Purification:
- Protein G purified
- Protein G purified
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Storage Buffer:
- Tris-Glycine and 0.15M NaCl
- Tris-Glycine and 0.15M NaCl
- Storage Temperature:
- Store at 4 °C short term. Aliquot and store at −20 °C long term. Avoid freeze-thaw cycles.
- Store at 4 °C short term. Aliquot and store at −20 °C long term. Avoid freeze-thaw cycles.