Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Undifferentiated Embryonic Cell transcription Factor 1
- Antigen Symbol:UTF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Epitope:C-PSPPKSADASPAP
- Gene ID:8433
- Antigen Synonyms:undifferentiated embryonic cell transcription factor 1
- Storage Buffer:0.5 mg/ml Tris (pH 7.3) and 0.5% BSA
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:Peptide with sequence C-PSPPKSADASPAP corresponding to internal region according to NP_003568.2.
- Tested Applications:Peptide
- Purification:Immunogen affinity purified
- Cat. No.:102182-382
- Supplier no.:NBP1-51943
Specifications
About this item
The UTF1 Antibody from Novus Biologicals is a goat polyclonal antibody to UTF1. This antibody reacts with human. The UTF1 Antibody has been validated for the following applications: Western Blot, Peptide ELISA.
Type: Primary
Antigen: UTF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-PSPPKSADASPAP
Host: Goat
Isotype: IgG
Reactivity: Human