Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:Scavenger Receptor Class B, Member 1
- Antigen Symbol:SR-BI
- Clonality:Polyclonal
- Conjugation:Biotin
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Hamster,Squirrel,Mustelid,Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μl
- Gene ID:949
- Antigen Synonyms:Collagen type I receptor|SCARB1|SR-B1|CD36 and LIMPII analogous 1|SRBI|SRB1 scavenger receptor class B member 1|CLA1 CD36 antigen-like 1|scavenger receptor class B type III|CLA-1 HDLQTL6|scavenger receptor class B|CD36L1scavenger receptor class B type 1|member 1|thrombospondin receptor)-like 1|CD36 antigen|thrombospondin receptor-like 1|CD36 antigen (collagen type I receptor|MGC138242
- Storage Buffer:PBS
- Storage Temperature:Store at 4C in the dark.
- Immunogen:A synthetic peptide made to the C-terminal region of mouse SR-BI (between residues 400-509). [UniProt# Q61009]
- Purification:Immunogen affinity purified
- Cat. No.:102131-478
- Supplier no.:NB400-131B
The SR-BI Antibody [Biotin] from Novus Biologicals is a goat polyclonal antibody to SR-BI. This antibody reacts with human, mouse, rat, hamster, mustelid, squirrel. The SR-BI Antibody [Biotin] has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Type: Primary
Antigen: SR-BI
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat, Hamster, Mustelid, Squirrel