Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Sodium Potassium ATPase Beta 1
- Antigen Symbol:ATP1B1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western Blot:Yes
- Size:100 μg
- Epitope:C-KTEISFRPNDPKSYE
- Gene ID:481
- Antigen Synonyms:ATP1B|sodium/potassium-transporting ATPase subunit beta-1|beta 1 polypeptide|Sodium/potassium-dependent ATPase subunit beta-1|K-ATPase beta-1 polypeptide|MGC1798|sodium/potassium-transporting ATPase beta-1 chain|sodium/potassium-dependent ATPase beta-1 subunit|adenosinetriphosphatase|K(+)-ATPase|Na|ATPase|Na+/K+ transporting|Beta 1-subunit of Na(+)
- Storage Buffer:0.5 mg/ml Tris (pH 7.3) and 0.5% BSA
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:Peptide with sequence C-KTEISFRPNDPKSYE corresponding to internal region according to NP_001668.1, NP_001001787.1.
- Tested Applications:Peptide
- Purification:Immunogen affinity purified
- Cat. No.:102182-468
- Supplier no.:NBP1-51987
Specifications
About this item
The Sodium Potassium ATPase Beta 1 Antibody from Novus Biologicals is a goat polyclonal antibody to Sodium Potassium ATPase Beta 1. This antibody reacts with mouse, rat. The Sodium Potassium ATPase Beta 1 Antibody has been validated for the following applications: Western Blot, Peptide ELISA.
Type: Primary
Antigen: Sodium Potassium ATPase Beta 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-KTEISFRPNDPKSYE
Host: Goat
Isotype: IgG
Reactivity: Mouse, Rat