Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:RAD9 Checkpoint Clamp Component A
- Antigen Symbol:Rad9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:5883
- Antigen Synonyms:cell cycle checkpoint control protein RAD9A|EC 3.1.11.2|RAD9 (S. pombe) homolog|RAD9 homolog A (S. pombe)|RAD9|hRAD9|DNA repair exonuclease rad9 homolog A
- Storage Buffer:0.5 mg/ml Tris (pH 7.3) and 0.5% BSA
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:Peptide with sequence C-QGPSPVLAEDSEGE corresponding to C-Terminus according to NP_004575.1.
- Tested Applications:Peptide Enzyme Immunoassay,
- Purification:Immunogen affinity purified
- Cat. No.:102110-322
- Supplier no.:NB100-57077
The Rad9 Antibody from Novus Biologicals is a goat polyclonal antibody to Rad9. This antibody reacts with human. The Rad9 Antibody has been validated for the following applications: Western Blot, Peptide ELISA, Enzyme Immunoassay, Sandwich ELISA.
Type: Primary
Antigen: Rad9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human