Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Purinergic Receptor P2X, Ligand-gated Ion channel 4
- Antigen Symbol:P2X4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μg
- Epitope:C Terminus
- Gene ID:5025
- Antigen Synonyms:purinergic receptor P2X|Purinergic receptor|P2X purinoceptor 4|ATP-gated cation channel protein|ATP receptor|purinoceptor P2X4|ligand-gated ion channel|subunit 4|4|P2X4P2X receptor|P2X4R
- Storage Buffer:0.5 mg/ml Tris (pH 7.3) and 0.5% BSA
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:Peptide with sequence C-YREKKYKYVEDYEQ corresponding to C-Terminus according to NP_002551.2.
- Tested Applications:Peptide
- Purification:Immunogen affinity purified
- Cat. No.:102154-594
- Supplier no.:NBP1-00141
Specifications
About this item
The P2X4 Antibody from Novus Biologicals is a goat polyclonal antibody to P2X4. This antibody reacts with human, mouse. The P2X4 Antibody has been validated for the following applications: Western Blot, Peptide ELISA.
Type: Primary
Antigen: P2X4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C Terminus
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse