Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Aldehyde Dehydrogenase 9 Family, member A1
- Antigen Symbol:ALDH9A1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Epitope:C-QKEILDKFTEEVVKQ
- Gene ID:223
- Antigen Synonyms:R-aminobutyraldehyde dehydrogenase|E34-trimethylaminobutyraldehyde dehydrogenase|member A1|EC 1.2.1.19|EC 1.2.1.3|EC 1.2.1|ALDH4|Aldehyde dehydrogenase family 9 member A1|aldehyde dehydrogenase 9 family|EC 1.2.1.47|ALDH7|Aldehyde dehydrogenase E3 isozyme|TMABADH|Gamma-aminobutyraldehyde dehydrogenase|ALDH9aldehyde dehydrogenase (NAD+)|EC 1.2.1.8
- Storage Buffer:0.5 mg/ml Tris (pH 7.3) and 0.5% BSA
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:Peptide with sequence C-QKEILDKFTEEVVKQ corresponding to internal region according to NP_000687.3.
- Purification:Immunogen affinity purified
- Cat. No.:102182-596
- Supplier no.:NBP1-52055
Specifications
About this item
The ALDH9A1 Antibody from Novus Biologicals is a goat polyclonal antibody to ALDH9A1. This antibody reacts with human. The ALDH9A1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Type: Primary
Antigen: ALDH9A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-QKEILDKFTEEVVKQ
Host: Goat
Isotype: IgG
Reactivity: Human