Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Transmembrane emp24 domain-containing protein 10
- Antigen Symbol:Tmp21/p23
- Clonality:Polyclonal
- Conjugation:DyLight® 650
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Primates,Rabbit,Chicken,Human,Bovine,Rat,Mouse
- Western Blot:Yes
- Size:100 μl
- Gene ID:10972
- Antigen Synonyms:transmembrane emp24 domain-containing protein 10|Transmembrane protein Tmp21,21 kDa transmembrane trafficking protein|S31III125|p23|S31I125|TMP21Tmp-21-I|transmembrane emp24-like trafficking protein 10 (yeast)|p24delta|P24(DELTA)|21 kDa transmembrane-trafficking protein
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4C in the dark.
- Immunogen:A synthetic peptide made to an internal sequence of the human TMP21 protein (within residues 100-200). [Swiss-Prot# P49755]
- Purification:Immunogen affinity purified
- Cat. No.:102123-400
- Supplier no.:NB110-57586C
Specifications
About this item
The Tmp21 / p23 Antibody [DyLight 650] from Novus Biologicals is a rabbit polyclonal antibody to Tmp21 / p23. This antibody reacts with human, mouse, rat, bovine, chicken, primate, rabbit. The Tmp21 / p23 Antibody [DyLight 650] has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: Tmp21/p23
Clonality: Polyclonal
Clone:
Conjugation: DyLight 650
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine, Chicken, Primate, Rabbit