Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:Urotensin-II R/GPR14
- Antigen Symbol:GPCR14
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Monkey,Human
- Size:0.05 ml
- Gene ID:2837
- Antigen Synonyms:UR-II-R|UR-2-R|urotensin 2 receptor|urotensin-2 receptor|GPR14|G protein-coupled receptor 14|UTR2|urotensin II receptor|G-protein coupled receptor 14|UTR
- Storage Buffer:PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:1.0 mg/ml
- Immunogen:Synthetic 20 amino acid peptide from N-terminal extracellular domain of human UTS2R/ GPR14.
- Purification:Immunogen affinity purified
- Cat. No.:102253-294
- Supplier no.:NLS372
The Urotensin-II R / GPR14 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Urotensin-II R / GPR14. This antibody reacts with human, monkey. The Urotensin-II R / GPR14 Antibody has been validated for the following applications: Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 3-13 ?g/ml
Type: Primary
Antigen: Urotensin-II R/GPR14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey