The Glut1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Glut1. This antibody reacts with human, mouse, rat. The Glut1 Antibody has been validated for the following applications: Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:100-1:250
Type: Primary
Antigen: Glut1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 102121-824
- 102121-812
- Antigen Symbol:
- Glut1
- Glut1
- Antigen Name:
- Glucose Transporter 1
- Glucose Transporter 1
- Antigen Synonyms:
- DYT18DYT17|erythrocyte/brain|GLUT1DS|HepG2 glucose transporter|facilitated glucose transporter member 1|MGC141896|GLUT-1|PED|member 1|solute carrier family 2 (facilitated glucose transporter)|GLUT1GLUT|solute carrier family 2|MGC141895|Glucose transporter type 1
- DYT18DYT17|erythrocyte/brain|GLUT1DS|HepG2 glucose transporter|facilitated glucose transporter member 1|MGC141896|GLUT-1|PED|member 1|solute carrier family 2 (facilitated glucose transporter)|GLUT1GLUT|solute carrier family 2|MGC141895|Glucose transporter type 1
- Antibody Type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 6513
- 6513
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- A synthetic peptide made to an N-terminal region of the human GLUT1 protein (between residues 1-100). [Swiss-Prot# P11166]
- A synthetic peptide made to an N-terminal region of the human GLUT1 protein (between residues 1-100). [Swiss-Prot# P11166]
- Purification:
- Immunogen affinity purified
- Immunogen affinity purified
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Storage Buffer:
- PBS
- PBS
- Storage Temperature:
- Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.