Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:FGF basic
- Antigen Symbol:FGF2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Horse,Human,Bovine,Rat
- Western Blot:Yes
- Size:100 μl
- Form:Lyophilized
- Gene ID:2247
- Antigen Synonyms:HBGF-2|FGFBprostatropin|bFGF|basic fibroblast growth factor bFGF|Basic fibroblast growth factor|fibroblast growth factor 2 (basic)|heparin-binding growth factor 2
- Storage Buffer:0.2 um filtered solution in PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:Synthetic peptide conjugated to KLH, corresponding to amino acids 1-24 of Cow FGF2.
- Purification:Sterile filtered
- Cat. No.:102133-464
- Supplier no.:NB600-1536
Specifications
About this item
The FGF basic / FGF2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to FGF basic / FGF2. This antibody reacts with human, rat, bovine, equine. The FGF basic / FGF2 Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: FGF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat, Bovine, Equine