Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:DEAH (Asp-Glu-Ala-His) box helicase 30
- Antigen Symbol:DHX30
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:22907
- Antigen Synonyms:ATP-dependent RNA helicase DHX30|FLJ11214|DEAH box protein 30|DDX30|KIAA0890RETCOR|DEAH (Asp-Glu-Ala-His) box polypeptide 30|EC 3.6.4.13|DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 30|retina co-repressor|putative ATP-dependent RNA helicase DHX30|EC 3.6.1
- Storage Buffer:TBS and 0.1% BSA
- Storage Temperature:Store at 4C. Do not freeze.
- Concentration:0.2 mg/ml
- Immunogen:The immunogen recognized by this antibody maps to a region between residue 1144 and 1194 of human DEAH (Asp-Glu-Ala-His) box polypeptide 30 using the numbering given in entry NP_619520.1 (GeneID 22907).
- Purification:Immunogen affinity purified
- Cat. No.:102161-066
- Supplier no.:NBP1-26604
Specifications
About this item
The DHX30 Antibody from Novus Biologicals is a rabbit polyclonal antibody to DHX30. This antibody reacts with human. The DHX30 Antibody has been validated for the following applications: Western Blot, Immunoprecipitation.
Type: Primary
Antigen: DHX30
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human