Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:Collagen XXV alpha 1
- Antigen Symbol:Collagen XXV
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μg
- Gene ID:84570
- Antigen Synonyms:alpha 1|collagen|AMY|Collagen-25|collagen alpha-1(XXV) chain|CLACP|CLAC-P|collagen-like Alzheimer amyloid plaque component|type XXV|collagenous Alzheimer amyloid plaque component|Alzheimer disease amyloid-associated protein|Collagen 25|CLAC
- Storage Buffer:PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:A synthetic peptide mapping to residues 155-169 [KGEQGDQGPRMVFPK] of the NC2-2 region of human and mouse CLAC-P.
- Purification:Immunogen affinity purified
- Cat. No.:102130-372
- Supplier no.:NB300-248
The Collagen XXV alpha 1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Collagen XXV alpha 1. This antibody reacts with human, mouse. The Collagen XXV alpha 1 Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: Collagen XXV alpha 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse