The ABCA1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to ABCA1. This antibody reacts with human, mouse, rat, porcine, canine, chicken, hamster, mustelid. The ABCA1 Antibody has been validated for the following applications: Western Blot, Simple Western, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Gel Supershift Assay.
Simple Western
Type: Primary
Antigen: ABCA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Porcine, Canine, Chicken, Hamster, Mustelid
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Catalog No:
- 102131-326
- 102131-340
- Antigen Symbol:
- ABCA1
- ABCA1
- Antigen Name:
- ATP Binding Cassette, sub-family A, member 1
- ATP Binding Cassette, sub-family A, member 1
- Antigen Synonyms:
- sub-family A (ABC1)|ABC-1|MGC165011|TGDATP-binding cassette sub-family A member 1|EC 3.6.3.41|MGC164864|ATP-binding cassette transporter 1|ABC1ATP-binding cassette transporter A1|ATP-binding cassette 1|CERP|HDLDT1|Cholesterol efflux regulatory protein|EC 3.6.3|FLJ14958|EC 2.7.7.8|member 1|membrane-bound|ATP-binding cassette|EC 2.8.1.8
- sub-family A (ABC1)|ABC-1|MGC165011|TGDATP-binding cassette sub-family A member 1|EC 3.6.3.41|MGC164864|ATP-binding cassette transporter 1|ABC1ATP-binding cassette transporter A1|ATP-binding cassette 1|CERP|HDLDT1|Cholesterol efflux regulatory protein|EC 3.6.3|FLJ14958|EC 2.7.7.8|member 1|membrane-bound|ATP-binding cassette|EC 2.8.1.8
- Antibody Type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Hamster, Pig, Mustelid, Chicken, Human, Dog, Rat, Mouse
- Hamster, Pig, Mustelid, Chicken, Human, Dog, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 19
- 19
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Gel Shift:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- Partial peptide sequence (within residues 1100-1300) of human ABCA1 [UniProt# O95477]. Actual immunogen sequence is proprietary information.
- Partial peptide sequence (within residues 1100-1300) of human ABCA1 [UniProt# O95477]. Actual immunogen sequence is proprietary information.
- Purification:
- Immunogen affinity purified
- Immunogen affinity purified
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Storage Buffer:
- PBS
- PBS
- Storage Temperature:
- Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.