About this item
The Ubiquitin Antibody (Ubi-1) from Novus Biologicals is a mouse monoclonal antibody to Ubiquitin. This antibody reacts with human, mouse, rat, bovine, c. elegans, chicken, drosophila, plant. The Ubiquitin Antibody (Ubi-1) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:500-1:1000
Type: Primary
Antigen: Ubiquitin
Clonality: Monoclonal
Clone: Ubi-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat, Bovine, C. elegans, Chicken, Drosophila, Plant
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 102129-970
- 102129-972
- Antigen Symbol:
- Ubi-1
- Ubi-1
- Antigen Name:
- Ubiquitin
- Ubiquitin
- Antigen Synonyms:
- UBB ubiquitin B|RPS27A|UBA52|UBC
- UBB ubiquitin B|RPS27A|UBA52|UBC
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- Ubi-1
- Ubi-1
- Reactivity:
- Drosophila, Plant, Chicken, C. elegans, Human, Bovine, Rat, Mouse
- Drosophila, Plant, Chicken, C. elegans, Human, Bovine, Rat, Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- 6233
- 6233
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Purified Ubiquitin conjugated to KLH with glutaraldehyde.
- Purified Ubiquitin conjugated to KLH with glutaraldehyde.
- Purification:
- Affinity purified
- Affinity purified
- Storage Buffer:
- Ascites
- Ascites
- Storage Temperature:
- Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.