Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Sodium/hydrogen exchanger 8
- Antigen Symbol:NHE8
- Clonality:Monoclonal
- Clone:7A11
- Conjugation:DyLight® 488
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μl
- Gene ID:23315
- Antigen Synonyms:member 8|KIAA0939DKFZp686C03237|MGC138418|NHE8FLJ42500|DKFZp686C03237|Solute carrier family 9 member 8|NHE-8|solute carrier family 9 (sodium/hydrogen exchanger)|sodium/hydrogen exchanger 8|Na(+)/H(+) exchanger 8|isoform 8
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4C in the dark.
- Immunogen:A fusion protein containing the C-terminal 89 residues of human NHE8. [Swiss-Prot# Q9Y2E8]
- Purification:Protein G purified
- Cat. No.:102124-562
- Supplier no.:NB110-62091G
Specifications
About this item
The NHE8 Antibody (7A11) [DyLight 488] from Novus Biologicals is a mouse monoclonal antibody to NHE8. This antibody reacts with human, mouse, rat. The NHE8 Antibody (7A11) [DyLight 488] has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: NHE8
Clonality: Monoclonal
Clone: 7A11
Conjugation: DyLight 488
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse, Rat