Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:Neuropeptide Y
- Antigen Symbol:NPY
- Clonality:Monoclonal
- Clone:8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human,Rat,Mouse
- Size:100 μg
- Epitope:The epitope of this antibody is probably around the mid-molecular region, NPY (18-24).
- Gene ID:4852
- Antigen Synonyms:170 kDa melanoma membrane-bound gelatinase|fibroblast activation protein|EC 3.4.21.-|Fibroblast activation protein alpha|alpha|seprase|DKFZp686G13158|DPPIV|Integral membrane serine protease|FAPA
- Storage Buffer:10mM Phosphate (pH 7.4) and 0.5M NaCl
- Storage Temperature:Store at 4C. Do not freeze.
- Concentration:1.0 mg/ml
- Immunogen:Synthetic human/rat NPY coupled to carrier with glutaraldehyde
- Purification:Protein A or G purified
- Cat. No.:102156-632
- Supplier no.:NBP1-05166
The Neuropeptide Y Antibody (8) - BSA Free from Novus Biologicals is a mouse monoclonal antibody to Neuropeptide Y. This antibody reacts with human, mouse, rat. The Neuropeptide Y Antibody (8) - BSA Free has been validated for the following applications: ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: Neuropeptide Y
Clonality: Monoclonal
Clone: 8
Conjugation: Unconjugated
Epitope: The epitope of this antibody is probably around the mid-molecular region, NPY (18-24).
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat