Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Calmodulin
- Antigen Symbol:CALM
- Clonality:Monoclonal
- Clone:2D1
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Bacteria,Chicken,Human,Bovine,Rat,Mouse
- Western Blot:Yes
- Size:200μl
- Gene ID:801
- Antigen Synonyms:calmodulin|delta subunit|CALM3|PHKDCAM|calmodulin 1 (phosphorylase kinase|CAM2|CAMB|CALML2|CALM2|delta)|CAMIII|DD132|CAM3|CAMC|CaM|CAMI|CAM1|phosphorylase kinase|CALM
- Storage Buffer:PBS
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Calmodulin purified from Dictyostelium discoideum.
- Purification:Protein A purified
- Cat. No.:102127-650
- Supplier no.:NB120-2860
Specifications
About this item
The Calmodulin Antibody (2D1) from Novus Biologicals is a mouse monoclonal antibody to Calmodulin. This antibody reacts with human, mouse, rat, bacteria, bovine, chicken. The Calmodulin Antibody (2D1) has been validated for the following applications: Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence, Immunocytochemistry.
Type: Primary
Antigen: CALM
Clonality: Monoclonal
Clone: 2D1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Bacteria, Bovine, Chicken