Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antigen Symbol:HIF-2 alpha/EPAS1
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:2 Vials
- Gene ID:2034
- Antigen Synonyms:Basic-helix-loop-helix-PAS protein MOP2|HIF2A|HIF2AHIF-2-alpha|Class E basic helix-loop-helix protein 73|hif2a angiogenesis|HIF-1alpha-like factor|bHLHe73ECYT4|endothelial PAS domain-containing protein 1|HLFHIF-1-alpha-like factor|EPAS-1|HIF-2 alpha|hypoxia-inducible factor 2 alpha|Member of PAS protein 2|MOP2HIF2-alpha|endothelial PAS domain protein 1|BHLHE73|PASD2PAS domain-containing protein 2|Hypoxia-inducible factor 2-alpha
- Storage Temperature:Storage is content dependent.
- Immunogen:NB 100-122 = Proprietary peptide derived from the mouse/human HIF-2a. NB 100-132 = Human HIF-2a amino acids 535-631.
- Cat. No.:102118-918
- Supplier no.:NB100-902HIF2
Mouse HIF-2 alpha Antibody Pack. Tested Applications: Western Blot, ELISA, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Frozen, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse, Rat.
Type:
Antigen: HIF2A
Clonality:
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human