Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:Slit Guidance Ligand 2
- Antigen Symbol:Slit2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 μg
- Gene ID:9353
- Antigen Synonyms:FLJ14420|slit (Drosophila) homolog 2|slit homolog 2 (Drosophila)|Slit-2SLIL3|slit homolog 2 protein
- Storage Buffer:0.5 mg/ml Tris (pH 7.3) and 0.5% BSA
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:Peptide with sequence DDCQDNKCKNGAH corresponding to internal region according to NP_004778.1.
- Tested Applications:, Peptide
- Purification:Immunogen affinity purified
- Cat. No.:102108-898
- Supplier no.:NB100-40807
The Slit2 Antibody from Novus Biologicals is a goat polyclonal antibody to Slit2. This antibody reacts with human. The Slit2 Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Recommended Dilutions: Immunohistochemistry (Parafin): 4-8 ?g/ml
Type: Primary
Antigen: Slit2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human