Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Solute Carrier Family 25 Member A47
- Antigen Symbol:SLC25A47
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 μg
- Epitope:KSRLQADGQGQRR
- Gene ID:283600
- Antigen Synonyms:solute carrier family 25|member 47|HDMCPsolute carrier family 25 member 47|Hepatocellular carcinoma down-regulated mitochondrial carrier protein|chromosome 14 open reading frame 68|C14orf68hepatocellular carcinoma-downregulated mitochondrial carrier protein
- Storage Buffer:0.5 mg/ml Tris (pH 7.3) and 0.5% BSA
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:Peptide with sequence C-KSRLQADGQGQRR corresponding to internal region according to NP_997000.2.
- Tested Applications:, Peptide
- Purification:Immunogen affinity purified
- Cat. No.:102110-354
- Supplier no.:NB100-57099
Specifications
About this item
The SLC25A47 Antibody from Novus Biologicals is a goat polyclonal antibody to SLC25A47. This antibody reacts with human. The SLC25A47 Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Type: Primary
Antigen: SLC25A47
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: KSRLQADGQGQRR
Host: Goat
Isotype: IgG
Reactivity: Human