Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Karyopherin Alpha 3
- Antigen Symbol:KPNA3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μg
- Gene ID:3839
- Antigen Synonyms:QIP2|SRP1-gamma|importin alpha 4|importin alpha-3|IPOA4|SRP1|SRP4|SRP1gamma|hSRP1|Importin alpha Q2|Karyopherin subunit alpha-3|importin-alpha-Q2|karyopherin alpha 3 (importin alpha 4)|importin subunit alpha-3
- Storage Buffer:0.5 mg/ml Tris (pH 7.3) and 0.5% BSA
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:Peptide with sequence C-DPTANLQTKEFNF corresponding to C-Terminus according to NP_002258.2.
- Purification:Immunogen affinity purified
- Cat. No.:102105-128
- Supplier no.:NB100-1373
Specifications
About this item
Goat Polyclonal KPNA3 Antibody. Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoprecipitation, Peptide ELISA. Tested Reactivity: Human, Mouse, Rat.
Type: Primary
Antigen: KPNA3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human