Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:BromoDomain Adjacent To Zinc Finger Domain, 2B
- Antigen Symbol:BAZ2B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 μg
- Gene ID:29994
- Antigen Synonyms:bromodomain adjacent to zinc finger domain|2B|FLJ45644|WALp4|KIAA1476|bromodomain adjacent to zinc finger domain protein 2B|DKFZp434H071|hWALp4|DKFZp762I0516
- Storage Buffer:0.5 mg/ml Tris (pH 7.3) and 0.5% BSA
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:Peptide with sequence C-YFEKKWTDTFKVS corresponding to C-Terminus according to NP_038478.2.
- Purification:Immunogen affinity purified
- Cat. No.:102104-688
- Supplier no.:NB100-1155
The BAZ2B Antibody from Novus Biologicals is a goat polyclonal antibody to BAZ2B. This antibody reacts with human. The BAZ2B Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Type: Primary
Antigen: BAZ2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human