Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:Autoimmune Regulator/AIRE
- Antigen Symbol:AIRE
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Pig,Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:326
- Antigen Synonyms:autoimmune regulator|autoimmune regulator (autoimmune polyendocrinopathy candidiasis ectodermaldystrophy)|APECED protein|Autoimmune polyendocrinopathy candidiasis ectodermal dystrophy protein|PGA1|autoimmune regulator (APECED protein)10APS1AIRE1|APECEDAPSI
- Storage Buffer:0.5 mg/ml Tris (pH 7.3) and 0.5% BSA
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:Peptide with sequence C-KDVDLSQPRKGRKP corresponding to internal region according to NP_000374.1.
- Purification:Immunogen affinity purified
- Cat. No.:102108-874
- Supplier no.:NB100-40799
The Autoimmune Regulator / AIRE Antibody from Novus Biologicals is a goat polyclonal antibody to Autoimmune Regulator / AIRE. This antibody reacts with human, porcine. The Autoimmune Regulator / AIRE Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Type: Primary
Antigen: Autoimmune Regulator/AIRE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Porcine