Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Meprin beta Subunit
- Antigen Symbol:MEP1B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Chicken
- Reactivity:Human
- Western Blot:Yes
- Size:0.05 mg
- Gene ID:4225
- Antigen Synonyms:endopeptidase-2|meprin A subunit beta|N-benzoyl-L-tyrosyl-P-amino-benzoic acid hydrolase subunit beta|meprin A|beta|Meprin B|EC 3.4.24.63|PPH beta|PABA peptide hydrolase|N-benzoyl-L-tyrosyl-p-amino-benzoic acid hydrolase beta subunit
- Storage Buffer:PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:1.0 mg/ml
- Immunogen:The immunogen for this antibody was a sequence from 379a.a. to 527a.a. from Meprin A beta(a 700a.a. long protein)
- Purification:Immunogen affinity purified
- Cat. No.:102116-696
- Supplier no.:NB100-75322
Specifications
About this item
Chicken Polyclonal Meprin beta Antibody. Tested Applications: Western Blot. Tested Reactivity: Human.
Type: Primary
Antigen: Meprin beta
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Chicken
Isotype:
Reactivity: Human