10x EDTA buffer pH 8.0, Novus Biologicals
Catalog # 102136-958
Supplier: Novus Biologicals
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Size:500 ml
- Storage Temperature:RT
- Cat. No.:102136-958
- Supplier no.:NB900-66730
Specifications
About this item
10x EDTA buffer pH 8.0.
- No preservative
Applications: Immunohistochemistry, Immunohistochemistry-Paraffin
Caution: This product is for research use only and is not approved for use in humans or in clinical diagnosis.