2-Propanol ≥99.5%, Cleanroom® LP for the electronics industry
Catalog # KM431-200440
Supplier: KMG
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Pack type:Poly bottle
- Size:1 gal.
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental Certification:EPA safer chemical solvent (Green circle rating); Chem21 solvent selection guide recommended by default and after discussion
- Cat. No.:KM431-200440
- Supplier no.:431-200440
Specifications
Specification Test Results
- Assay (C3H8O):99.5% Min.
- Assay (H2O):500 ppm
- Color:10 APHA
- Appearance:Conforms
- Acidity:0.200 μeq/g
- Alkalinity:0.100 μeq/g
- Residue After Evaporation:4,000 ppb
- Solubility (in H2O):Conforms
- Infrared:Conforms
- Chloride (Cl):100 ppb
- Phosphate (PO4):300 ppb
- Aluminum (Al):50.0 ppb
- Antimony (Sb):5.0 ppb
- Arsenic (As):5.0 ppb
- Barium (Ba):100.0 ppb
- Beryllium (Be):100.0 ppb
- Bismuth (Bi):100.0 ppb
- Boron (B):10.0 ppb
- Cadmium (Cd):100.0 ppb
- Calcium (Ca):50.0 ppb
- Chromium (Cr):50.0 ppb
- Cobalt (Co):50.0 ppb
- Copper (Cu):10.0 ppb
- Gallium (Ga):50.0 ppb
- Germanium (Ge):50.0 ppb
- Gold (Au):50.0 ppb
- Iron (Fe):50.0 ppb
- Lead (Pb):50.0 ppb
- Lithium (Li):50.0 ppb
- Magnesium (Mg):50.0 ppb
- Manganese (Mn):15.0 ppb
- Molybdenum (Mo):100.0 ppb
- Nickel (Ni):10.0 ppb
- Niobium (Nb):100.0 ppb
- Potassium (K):75.0 ppb
- Silicon (Si):100.0 ppb
- Silver (Ag):50.0 ppb
- Sodium (Na):75.0 ppb
- Strontium (Sr):100.0 ppb
- Tantalum (Ta):100.0 ppb
- Thallium (Tl):100.0 ppb
- Tin (Sn):50.0 ppb
- Titanium (Ti):50.0 ppb
- Vanadium (V):100.0 ppb
- Zinc (Zn):50.0 ppb
- Zirconium (Zr):100.0 ppb
- 0.2μ Particle Count:1,000 par/ml
- 0.3μ Particle Count:500 par/ml
- 0.5μ Particle Count:50 par/ml
- 1.0μ Particle Count:8 par/ml
Specification Test Results
About this item
Synonyms: Isopropyl rubbing alcohol, sec-Propanol, Isopropanol, 2-Hydroxypropane, Isopropyl Rubbing Alcohol, 2-Hydroxy propane, IPA, sec-Propyl Alcohol, Dimethyl carbinol, Propane, 2-hydroxy-, Isopropyl alcohol, Propan-2-ol
:
:60.1 g/mol
Flash Pt:12
EINECS:200-661-7
UN:1219
REACH:01-2119457558-25
:GHS02
:GHS07
Storage Temperature:Ambient
CAS Number:67-63-0
Formula:(CH₃)₂CHOH
MDL Number:MFCD00011674
Boiling Pt:82
Melting Pt:-89
Density:0.786 g/cm³ (20 °C)
ADR:3 II
Merck Index:13,05228