Order Entry
United States
ContactUsLinkComponent
E.Z.N.A.® Plasmid Midi Kit, Omega Bio-tek®
E.Z.N.A.® Plasmid Midi Kit, Omega Bio-tek®
Supplier:  Omega Bio-Tek
Certificates
E.Z.N.A.® Plasmid Midi Kit, Omega Bio-tek®
Supplier:  Omega Bio-Tek

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Midi
  • Refine target molecule:
    Plasmid
  • Sample type:
    Bacteria
  • Target molecule:
    Plasmid
  • Description:
    Plasmid Midi Kit

Specifications

Frequently Bought Together