Corning® HBSS (Hanks Balanced Salt Solution), Corning
Supplier: Corning
Certificates
About this item
Hanks' Balanced Salt Solution (HBSS) is a buffer used to maintain a physiological pH for cells maintained in non-CO₂ atmospheric conditions.
- Low endotoxin - < 0.25EU/mL
- Mycoplasma tested
- Liquid media products are formulated with cell culture grade WFI water
- Manufactured in a cGMP environment
Specifications
- Mycoplasma:Tested Negative
- Endotoxin Content:< 0.25 EU/mL
- Shelf Life:36 months
- Storage Temperature:2 °C to 8 °C
Specifications
Related Information
Certificates
Search by Lot Number3D Cell Culture Quick Reference Ordering Guide Corning 3D Cell Culture Quick Reference Ordering Guide Corning Cell Culture Media Quick Reference Guide Updated.pdf Corning® Shopping list for returning to the bench
Product Literature
Cell Culture Media, Molecular Buffers and Reagents
Additional Product Details
Corning Cell Culture Media.pdf
Promotion Material
Frequently Bought Together