Order Entry
United States
ContactUsLinkComponent
Precipitation Solution, OmniPur®
Precipitation Solution, OmniPur®
Catalog # EM-7280
Supplier:  MilliporeSigma
CAS Number:  
Precipitation Solution, OmniPur®
Catalog # EM-7280
Supplier:  MilliporeSigma
Supplier Number:  7280-20L
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    20 L
  • Cat. No.:
    EM-7280
  • Supplier no.:
    7280-20L

Specifications

Specification Test Results

  • Appearance:
    Clear, colorless liquid
  • Conductivity at 25°C (of a 1:1000 dilution):
    350-650µS
  • DNase:
    None detected
  • pH at 25°C:
    7.2-7.7
  • Protease:
    None detected
  • RNase:
    None detected
  • Titration (molarity):
    4.85-5.10M

Specification Test Results

About this item

 :