Order Entry
United States
ContactUsLinkComponent
Corning® Serum-Free/Low-Protein Medium, 1X with L-Glutamine and 1 g/L BSA, Corning
Corning® Serum-Free/Low-Protein Medium, 1X with L-Glutamine and 1 g/L BSA, Corning
Catalog # 45000-420
Supplier:  Corning
CAS Number:  
undefined
Corning® Serum-Free/Low-Protein Medium, 1X with L-Glutamine and 1 g/L BSA, Corning
Catalog # 45000-420
Supplier:  Corning
Supplier Number:  40-101-CV
CAS Number:  

Specifications

  • Media Format:
    Liquid
  • Description:
    Corning® cellgro® COMPLETE Serum-Free/Low-Protein Medium, 1X with L-Glutamine and 1 g/L BSA
  • Size:
    500 mL
  • Storage Temperature:
    Refrigerated
  • Shipping Temperature:
    Ambient
  • Cat. No.:
    45000-420
  • Supplier no.:
    40-101-CV

Specifications

About this item

Corning® cellgro® COMPLETE formulation is based on a proprietary combination of 1:1 DMEM/F12, RPMI and McCoy's 5A medium. This medium contains a mixture of selected trace elements and high molecular weight carbohydrates, extra vitamins, animal origin-free protein, and a small amount of high quality bovine serum albumin (1.0g/L). The Corning cellgro COMPLETE supports superior growth and viability over long-term passages in hybridomas, along with suspension and adherent cell cultures.