ContactUsLinkComponent
Corning® EDTA, 0.5M pH 8.0, Corning
Corning® EDTA, 0.5M pH 8.0, Corning
  45001-122
 :  Corning
 :  
undefined
Corning® EDTA, 0.5M pH 8.0, Corning
  45001-122
 :  Corning
 :  46-034-CI
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Pack type:
    Plastic bottle
  • Description:
    0.5M EDTA, pH 8.0
  • Size:
    100 mL
  • Storage Temperature:
    Ambient
  • Shipping Temperature:
    Ambient
  • Cat. No.:
    45001-122
  • Supplier no.:
    46-034-CI
EDTA (ethylenediaminetetraacetic acid) is a polyamino carboxylic acid. It is a colorless, water-soluble solid and is useful as a chelating agent due to its ability to "sequester" metal ions such as Ca2+ and Fe3+.  These cations are responsible for various undesirable effects in molecular biological and cell culture processes. After being bound by EDTA, these metal ions remain in the solution but exhibit diminished reactivity.  The use of EDTA therefore allows for greater control over many experiments.