Silver nitrate 0.01 N in isopropanol (IPA)
Catalog # RCSOD90-32
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Size:32 oz.
- Cat. No.:RCSOD90-32
- Supplier no.:RSOD9000-1C
Specifications
About this item
:
:169.87 g/mol
UN:1493
ADR:5.1,II
CAS Number:7761-88-8
Formula:AgNO₃
MDL Number:MFCD00003414