Acetonitrile : water : formic acid (95:5:0.1)%, LabReady™, Solvent Blend for LC-MS, for HPLC, Burdick & Jackson™
Catalog # BJLC599-2.5
Supplier: Honeywell Research Chemicals
Product available on GSA Advantage®
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:2.5 L
- Cat. No.:BJLC599-2.5
- Supplier no.:LC599-2.5
Specifications
About this item
:
Boiling Pt:70 °C
Density:0.7951 g/cm³ (20 °C)
Flash Pt:6.67 °C
Storage Temperature:Ambient
UN:1648
ADR:3,II
:GHS02
:GHS07
:GHS08
:7732-18-5